Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tributyl[(methoxymethoxy)methyl]stannane ≥94%

Supplier: MATRIX SCIENTIFIC MS

070168-1G 070168-5G 070168-25G
101943-626EA 553.85 USD InStock
101943-626 101943-628 101943-630
Tributyl[(methoxymethoxy)methyl]stannane ≥94%
Tributyl[(methoxymethoxy)methyl]stannane
Hazard: Irritant


Formula: C₁₅H₃₄O₂Sn
MDL Number: MFCD03412101
CAS Number: 100045-83-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty