Ethyl-4-amino-1-piperidinecarboxylate

Supplier: MATRIX SCIENTIFIC MS

037466-5G 037466-25G
101843-458EA 98.46 USD
101843-458 101843-460
Ethyl-4-amino-1-piperidinecarboxylate
Ethyl-4-amino-1-piperidinecarboxylate

Hazard: Irritant

Formula: C₈H₁₆N₂O₂
MW: 172.23 g/mol
Boiling Pt: 118…120 °C (6 mmHg)
Flash Pt: 78 °C
MDL Number: MFCD00006484
CAS Number: 58859-