Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-bromo-1-methyl-1H-indazole-3-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

068469-250MG 068469-500MG 068469-1G
101936-288EA 1076.92 USD InStock
101936-288 101936-290 101936-292
Methyl-6-bromo-1-methyl-1H-indazole-3-carboxylate ≥95%
Methyl-6-bromo-1-methyl-1H-indazole-3-carboxylate
Hazard: Irritant


Formula: C₁₀H₉BrN₂O₂
MDL Number: MFCD15071437
CAS Number: 946427-77-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty