Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl piperidin-1-ylcarbamate ≥95%

Supplier: MATRIX SCIENTIFIC MS

068994-250MG 068994-500MG 068994-1G
101938-576EA 290.77 USD InStock
101938-576 101938-578 101938-580
tert-Butyl piperidin-1-ylcarbamate ≥95%
tert-Butyl piperidin-1-ylcarbamate
Hazard: Irritant


Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD00961870
CAS Number: 126216-45-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty