Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4-Difluoro-1-(hydroxymethyl)adamantane ≥95%

Supplier: MATRIX SCIENTIFIC MS

070730-250MG 070730-500MG 070730-1G
101946-086EA 2907.69 USD InStock
101946-086 101946-088 101946-090
4,4-Difluoro-1-(hydroxymethyl)adamantane ≥95%
4,4-Difluoro-1-(hydroxymethyl)adamantane
Hazard: Irritant


Formula: C₁₁H₁₆F₂O
MDL Number: MFCD18800684
CAS Number: 1283719-51-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty