Glycerine ≥99.5%, Molecular Biology Grade

Supplier: Jopak Verpakkingen

IB15760 IB15762
IB15760EA 264.72 USD
IB15760 IB15762
Glycerine ≥99.5%, Molecular Biology Grade
Glycerine

Glycerol is used to prevent freezing during enzyme and bacterial preparations.


Dnase, Rnase, and Protease Free


Precaución: Store at room temperature, protect from moisture, and keep tightly sealed.

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=481dd90ecbdec73cd4020432d3356161ebfe7681&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity >99.5%
Sulfate <0.0001%
Arsenic <0.0003%
Chlorine <0.0003%
Iron <0.0005%
Residue on Ignition <0.01%
Density 1.249-1.262 g/ml
Specific Gravity 1.2635 – 1.2615
DNase Assay None Detected
RNase Assay None Detected
Protease Assay None Detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR