IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99.0%, Molecular Biology Grade
Supplier: Jopak Verpakkingen
IPTG is an inducer of β-galactosidase activity in E.coli. It is commonly used in conjunction with X-GAL to detect lac gene expression in cloning applications, thus allowing detection of recombinant molecules.
- Used to induce β-galactosidase activity in E. coli
Store dessicated at -20°C, keep tightly sealed, and warm to room temperature before opening.
Formula:
C₉H₁₈O₅S MW: 238.31 g/mol Boiling Pt: 402 |
MDL Number:
MFCD00063273 CAS Number: 367-93-1 |