Ehrlich aldehyde reagent, Wallace-diamond formulation for urobilinogen determination
Supplier: RICCA CHEMICAL CO
|
Danger
|
R2780000-500C
R2780000-120C
RC278016EA
90
USD
RC278016
RC27804
Ehrlich aldehyde reagent, Wallace-diamond formulation for urobilinogen determination
Ehrlich aldehyde reagent, Wallace-diamond formulation
Boiling Pt:
~ 100 °C Melting Pt: ~ 0 °C Density: 1.0500 g/cm³ Storage Temperature: Ambient |
UN:
1789 ADR: 8 II |
Specification Test Results
Appearance | Clear, greenish-yellow, slightly pungent to pungent odor |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...