Acetonitrile 99,5

Supplier: Decon Laboratories Ltd
Danger

A0060-25ML A0060-500ML
TCA0060-025MLEA 17 USD
TCA0060-025ML TCA0060-500ML
Acetonitrile 99,5
Acetonitrile

CAS Number: 75-05-8
MDL Number: MFCD00001878
Molecular Formula: C2H3N
Molecular Weight: 41.05
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Melting point (°C): -46
Flash Point (°C): 10
Specific Gravity (20/20): 0.78


Other Product Names / Synonyms: Methyl Cyanide, ACN

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Density: 0.78 (20 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=448a84cb7a60315bb069f40c31fefc6339139261&vid=448a84cb7a60315bb069f40c31fefc6339139261&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR