Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-1-hexyne ≥97.0%

Supplier: Decon Laboratories Ltd

C1493-5ML C1493-25ML
TCC1493-005MLEA 97 USD InStock
TCC1493-005ML TCC1493-25ML
6-Chloro-1-hexyne ≥97.0%
6-Chloro-1-hexyne
CAS Number: 10297-06-0
MDL Number: MFCD00013697
Molecular Formula: C6H9Cl
Molecular Weight: 116.59
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 34
Specific Gravity (20/20): 0.96

Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 5-Hexynyl Chloride


Formula: C₆H₉Cl
Boiling Pt: 144 °C
Density: 0.96 (20 °C)
Flash Pt: 34 °C
MDL Number: MFCD00013697
CAS Number: 10297-06-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4ba5d952a73d026ef0308a9917979ac93432e33&vid=e4ba5d952a73d026ef0308a9917979ac93432e33&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty