D2198-25G
D2198-5G
TCD2198-025GEA
256
USD
InStock
TCD2198-025G
TCD2198-5G
2,4-Dichlorobenzyl mercaptan ≥98.0%
2,4-Dichlorobenzyl mercaptan
CAS Number: 59293-67-3
MDL Number: MFCD00039650
Molecular Formula: C7H6Cl2S
Molecular Weight: 193.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Specific Gravity (20/20): 1.36
Other Product Names / Synonyms: 2,4-Dichloro-alpha-mercaptotoluene, 2,4-Dichlorothiobenzyl Alcohol, 2,4-Dichloro-alpha-toluenethiol, (2,4-Dichlorophenyl)methanethiol