Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxysalicylic acid ≥98.0%

Supplier: Decon Laboratories Ltd

M1254-25G M1254-5G
TCM1254-025GEA 105 USD InStock
TCM1254-025G TCM1254-5G
5-Methoxysalicylic acid ≥98.0%
5-Methoxysalicylic acid
CAS Number: 2612-02-4
MDL Number: MFCD00002459
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 144

Other Product Names / Synonyms: 2-Hydroxy-5-methoxybenzoic Acid


Formula: C₈H₈O₄
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 144 °C
Storage Temperature: Ambient
MDL Number: MFCD00002459
CAS Number: 2612-02-4
EINECS: 220-037-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0bdb2622130f8fa88d5b66e003adc56b72bf8cc&vid=b0bdb2622130f8fa88d5b66e003adc56b72bf8cc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty