Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Triacontanol ≥85.0%

Supplier: Decon Laboratories Ltd

T1049-100MG T1049-1G
TCT1049-100MGEA 155 USD InStock
TCT1049-100MG TCT1049-1G
1-Triacontanol ≥85.0%
1-Triacontanol
CAS Number: 593-50-0
MDL Number: MFCD00002963
Molecular Formula: C30H62O
Molecular Weight: 438.83
Purity/Analysis Method: >85.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 86


Formula: H₃C(CH₂)₂₈CH₂OH
Boiling Pt: 443 °C (1013 hPa)
Melting Pt: 86 °C
Density: 0.777 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002963
CAS Number: 593-50-0
EINECS: 209-794-5
Merck Index: 12,09722
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45a7efafb432c113edb13404c3b20dd2223dddc8&vid=45a7efafb432c113edb13404c3b20dd2223dddc8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >85.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty