Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloromethylstyrene (m and p mixture) ≥88.0% stabilized

Supplier: Decon Laboratories Ltd

C0767-500G C0767-25G
TCC0767-500GEA 462 USD InStock
TCC0767-500G TCC0767-025G
Chloromethylstyrene (m and p mixture) ≥88.0% stabilized
Chloromethylstyrene (m and p mixture)
(stabilized with TBC + ONP + o-Nitrocresol)
CAS Number: 57458-41-0
MDL Number: MFCD00051362
Molecular Formula: C9H9Cl
Molecular Weight: 152.62
Purity/Analysis Method: >88.0% (GC)
Form: Clear Liquid
Flash Point (°C): 107
Specific Gravity (20/20): 1.10
Storage Temperature: 0-10°C

Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Vinylbenzyl Chloride


Formula: C₉H₉Cl
Density: 1.10 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00051362
CAS Number: 57458-41-0
Order Now
Specification Test Results
Purity/Analysis Method >88.0% (GC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty