D1032-1G
D1032-100MG
TCD1032-001GEA
112.57
USD
InStock
TCD1032-001G
TCD1032-100MG
N-(2,4-Dinitrophenyl)glycine ≥98.0% (by HPLC, titration analysis)
N-(2,4-Dinitrophenyl)glycine
CAS Number: 1084-76-0
MDL Number: MFCD00024400
Molecular Formula: C8H7N3O6
Molecular Weight: 241.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 200
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Dnp-glycine, Dnp-Gly-OH