Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2,4-Dinitrophenyl)-L-proline ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

D1036-1G D1036-100MG
TCD1036-001GEA 144.4 USD InStock
TCD1036-001G TCD1036-100MG
N-(2,4-Dinitrophenyl)-L-proline ≥98.0% (by HPLC, titration analysis)
N-(2,4-Dinitrophenyl)-L-proline
CAS Number: 1655-55-6
MDL Number: MFCD00037319
Molecular Formula: C11H11N3O6
Molecular Weight: 281.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 140
Specific rotation [a]20/D: -682.5 deg (C=0.5, AcOH)

Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Dnp-L-proline, Dnp-Pro-OH


Formula: C₁₁H₁₁N₃O₆
Melting Pt: 140 °C
MDL Number: MFCD00037319
CAS Number: 1655-55-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a10f3f8a67d275763a91f90e34b13ede6f00591f&vid=a10f3f8a67d275763a91f90e34b13ede6f00591f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty