Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Dimethylamino)propanoic acid hydrochloride ≥98.0%

Supplier: Decon Laboratories Ltd

D2690-25G D2690-5G
TCD2690-25GEA 144 USD InStock
TCD2690-25G TCD2690-005G
3-(Dimethylamino)propanoic acid hydrochloride ≥98.0%
3-(Dimethylamino)propanoic acid hydrochloride
CAS Number: 14788-12-6
MDL Number: MFCD02093464
Molecular Formula: C5H11NO2
Molecular Weight: 153.61
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 189

Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N,N-Dimethyl-beta-alanine Hydrochloride


Formula: C5H12ClNO2
Melting Pt: 189 °C
MDL Number: MFCD02093464
CAS Number: 14788-12-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f47a377b2d767e4f8cb5621399cf6e4d84ad587&vid=0f47a377b2d767e4f8cb5621399cf6e4d84ad587&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty