H0002-1G
H0002-100MG
TCH0002-001GEA
119
USD
InStock
TCH0002-001G
TCH0002-100MG
Harmine hydrochloride ≥98.0% (by HPLC, titration analysis)
Harmine hydrochloride
CAS Number: 343-27-1
MDL Number: MFCD00012641
Molecular Formula: C13H12N2O
Molecular Weight: 248.71
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Indole Alkaloids
Other Product Names / Synonyms: Banisterine Hydrochloride