Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Isopropoxypropylamine ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

I0162-25ML I0162-500ML
TCI0162-025MLEA 32 USD InStock
TCI0162-025ML TCI0162-500ML
3-Isopropoxypropylamine ≥98.0% (by GC, titration analysis)
3-Isopropoxypropylamine
CAS Number: 2906-12-9
MDL Number: MFCD00008220
Molecular Formula: C6H15NO
Molecular Weight: 117.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 39
Specific Gravity (20/20): 0.84

Other Product Names / Synonyms: 3-Aminopropyl Isopropyl Ether


Formula: C₆H₁₅NO
Boiling Pt: 144 °C
Density: 0.84 (20 °C)
Flash Pt: 39 °C
MDL Number: MFCD00008220
CAS Number: 2906-12-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef06b8c5858921fd9486e664a931086e46d2461f&vid=ef06b8c5858921fd9486e664a931086e46d2461f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty