Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl-(S)-(-)-2-isocyanatoglutarate ≥97.0%

Supplier: Decon Laboratories Ltd

I0468-5G I0468-1G
TCI0468-005GEA 425.8 USD InStock
TCI0468-005G TCI0468-001G
Diethyl-(S)-(-)-2-isocyanatoglutarate ≥97.0%
Diethyl-(S)-(-)-2-isocyanatoglutarate
CAS Number: 145080-95-1
MDL Number: MFCD00191663
Molecular Formula: C10H15NO5
Molecular Weight: 229.23
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Specific Gravity (20/20): 1.13
Specific rotation [a]20/D: -43.5 deg (neat)
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: Diethyl (S)-(-)-2-Isocyanatopentanedioate, (S)-(-)-2-Isocyanatoglutaric Acid Diethyl Ester, (S)-(-)-2-Isocyanatopentanedioic Acid Diethyl Ester


Formula: C₁₀H₁₅NO₅
MW: 229.23 g/mol
Boiling Pt: 151 °C
Density: 1.13 (20 °C)
MDL Number: MFCD00191663
CAS Number: 145080-95-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2120db491b469d57af6f7df848003eca7d0d74e&vid=c2120db491b469d57af6f7df848003eca7d0d74e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty