Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methylthio)propionyl chloride ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

M0806-25ML M0806-500ML
TCM0806-025MLEA 118 USD InStock
TCM0806-025ML TCM0806-500ML
3-(Methylthio)propionyl chloride ≥98.0% (by titrimetric analysis)
3-(Methylthio)propionyl chloride
CAS Number: 7031-23-4
MDL Number: MFCD00059482
Molecular Formula: C4H7ClOS
Molecular Weight: 138.61
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 96
Specific Gravity (20/20): 1.20

Other Product Names / Synonyms: 3-(Methylmercapto)propionyl Chloride


Formula: C₄H₇ClOS
Boiling Pt: 96 °C
Density: 1.20 (20 °C)
MDL Number: MFCD00059482
CAS Number: 7031-23-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=796cfb7da7e3e2fb6cb39c8f6998ebaac6ee7e5b&vid=796cfb7da7e3e2fb6cb39c8f6998ebaac6ee7e5b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty