Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Mercapto-1-propanol ≥97.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

M1206-25G M1206-5G
TCM1206-025GEA 217 USD InStock
TCM1206-025G TCM1206-5G
3-Mercapto-1-propanol ≥97.0% (by GC, titration analysis)
3-Mercapto-1-propanol
CAS Number: 19721-22-3
MDL Number: MFCD00192262
Molecular Formula: C3H8OS
Molecular Weight: 92.16
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 85
Specific Gravity (20/20): 1.07


Formula: C₃H₈OS
Boiling Pt: 90 °C
Density: 1.07 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00192262
CAS Number: 19721-22-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=565c662e7a58e31c1b7f48e64cafc38dee3a4f39&vid=565c662e7a58e31c1b7f48e64cafc38dee3a4f39&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty