Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Nitrophenoxy)acetic acid ≥98.0% for synthesis

Supplier: Decon Laboratories Ltd

N0226-500G N0226-25G
TCN0226-500GEA 412 USD InStock
TCN0226-500G TCN0226-025G
(4-Nitrophenoxy)acetic acid ≥98.0% for synthesis
(4-Nitrophenoxy)acetic acid
CAS Number: 1798-11-4
MDL Number: MFCD00017030
Molecular Formula: C8H7NO5
Molecular Weight: 197.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 187


Formula: (C₆H₄(NO₂))OCH₂COOH
MW: 197.15 g/mol
Melting Pt: 187 °C
MDL Number: MFCD00017030
CAS Number: 1798-11-4
EINECS: 217-283-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ada7602283cb365130552f2bb504aee3aa4a201d&vid=ada7602283cb365130552f2bb504aee3aa4a201d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty