Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3',3''-Nitrilotripropionic acid ≥99.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

N0409-25G N0409-10G
TCN0409-025GEA 312 USD InStock
TCN0409-025G TCN0409-010G
3,3',3''-Nitrilotripropionic acid ≥99.0% (by titrimetric analysis)
3,3',3''-Nitrilotripropionic acid
CAS Number: 817-11-8
MDL Number: MFCD00055827
Molecular Formula: C9H15NO6
Molecular Weight: 233.22
Purity/Analysis Method: >99.0% (T)
Form: Crystal


Formula: C₉H₁₅NO₆
MDL Number: MFCD00055827
CAS Number: 817-11-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ba5b52c6339d0ce78d3af41c417d660c2495b77&vid=7ba5b52c6339d0ce78d3af41c417d660c2495b77&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty