P0619-5G
P0619-25G
TCP0619-005GEA
71
USD
InStock
TCP0619-005G
TCP0619-025G
Picolinoyl chloride hydrochloride ≥93.0%
Picolinoyl chloride hydrochloride
CAS Number: 39901-94-5
MDL Number: MFCD00060193
Molecular Formula: C6H4ClNO
Molecular Weight: 178.01
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Picolinoyl Chloride Hydrochloride