Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Lauroylsarcosine sodium salt hydrate ≥98.0% (by total nitrogen basis) for biochemical research

Supplier: Decon Laboratories Ltd

S0597-5G S0597-25G
TCS0597-5GEA 24 USD InStock
TCS0597-5G TCS0597-25G
N-Lauroylsarcosine sodium salt hydrate ≥98.0% (by total nitrogen basis) for biochemical research
N-Lauroylsarcosine sodium salt hydrate
[for Biochemical Research]
CAS Number: 137-16-6
MDL Number: MFCD00042728
Molecular Formula: C15H29NO3
Molecular Weight: 293.38
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 46

Product Categories: Bioscience, Anionic Surfactants (for Biochemistry)
Other Product Names / Synonyms: N-Lauroylsarcosine Sodium Salt, Sodium N-Dodecanoylsarcosinate, N-Dodecanoylsarcosine Sodium Salt


Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
Melting Pt: 46 °C
MDL Number: MFCD00042728
CAS Number: 137-16-6
UN: 2811
ADR: 6.1,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eecdaeea8eb1ed102e0bdf317e12a71c5006ecf8&vid=eecdaeea8eb1ed102e0bdf317e12a71c5006ecf8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty