T0611-25G
T0611-5G
TCT0611-025GEA
301
USD
InStock
TCT0611-025G
TCT0611-005G
Trichloromethanesulfonyl chloride ≥95.0% (by titrimetric analysis)
Trichloromethanesulfonyl chloride
CAS Number: 2547-61-7
MDL Number: MFCD00007452
Molecular Formula: CCl4O2S
Molecular Weight: 217.87
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 141