Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(dimethylamino)methane ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

T1057-5ML T1057-25ML
TCT1057-005MLEA 254 USD InStock
TCT1057-005ML TCT1057-25ML
Tris(dimethylamino)methane ≥97.0% (by titrimetric analysis)
Tris(dimethylamino)methane
CAS Number: 5762-56-1 MDL Number: MFCD00008322 Molecular Formula: C7H19N3 Molecular Weight: 145.25 Purity/Analysis Method: 97.0% (T) Form: Clear Liquid Color: Colorless Boiling point (°C): 43 Flash Point (°C): -15 Specific Gravity (20/20): 0.84


Formula: C₇H₁₉N₃
Boiling Pt: 43 °C
Density: 0.84 (20 °C)
Flash Pt: –15 °C
MDL Number: MFCD00008322
CAS Number: 5762-56-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcfdb8cd914b221b50ee63726e39521753f7e548&vid=dcfdb8cd914b221b50ee63726e39521753f7e548&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty