Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethylisopropylsilyl trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

T1468-1G T1468-5G
TCT1468-001GEA 111 USD InStock
TCT1468-001G TCT1468-5G
Diethylisopropylsilyl trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
Diethylisopropylsilyl trifluoromethanesulfonate
CAS Number: 126889-55-2
MDL Number: MFCD00191689
Molecular Formula: C8H17F3O3SSi
Molecular Weight: 278.36
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Diethylisopropylsilyl Triflate, Trifluoromethanesulfonic Acid Diethylisopropylsilyl Ester


Formula: C₈H₁₇F₃O₃SSi
MDL Number: MFCD00191689
CAS Number: 126889-55-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15020f6a32fb906bbebc0f0d02ed511f35f1de3d&vid=15020f6a32fb906bbebc0f0d02ed511f35f1de3d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty