Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,5-Trichlorobenzenesulfonic acid hydrate ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

B0248-25G B0248-5G
TCB0248-025GEA 267 USD InStock
TCB0248-025G TCB0248-5G
2,4,5-Trichlorobenzenesulfonic acid hydrate ≥98.0% (by titrimetric analysis)
2,4,5-Trichlorobenzenesulfonic acid hydrate
CAS Number: 6378-25-2 MDL Number: MFCD00070539 Molecular Formula: C6H3Cl3O3S Molecular Weight: 261.50 Purity/Analysis Method: 98.0% (T) Form: Crystal


Formula: C₆H₃Cl₃O₃S
MDL Number: MFCD00070539
CAS Number: 6378-25-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fd3f501dc07bf135db7d0b87cdb8e0b1d623283&vid=3fd3f501dc07bf135db7d0b87cdb8e0b1d623283&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty