Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4,4,4-trifluorocrotonate ≥95%

Supplier: MATRIX SCIENTIFIC MS

002412-25G 002412-5G
101776-058EA 590.77 USD InStock
101776-058 101776-056
Methyl-4,4,4-trifluorocrotonate ≥95%
Methyl 4,4,4-trifluorocrotonate
Hazard: Flammable/Irritant/Refrigerate


Formula: C₅H₅F₃O₂
Boiling Pt: 95 °C
Density: 1.237
Flash Pt: 32 °C
MDL Number: MFCD00077567
CAS Number: 85694-31-1
UN: 3272
ADR: 3,III
Order Now
Specification Test Results
Boiling point 95°
ND20 1.3639

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty