Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1,1,2,2-Tetrafluoroethoxy)benzene ≥98%

Supplier: MATRIX SCIENTIFIC MS

002871-5G 002871-25G
101776-404EA 196.92 USD InStock
101776-404 101776-406
(1,1,2,2-Tetrafluoroethoxy)benzene ≥98%
(1,1,2,2-Tetrafluoroethoxy)benzene
Hazard: Flammable


Formula: HF₂CCF₂OC₆H₅
Boiling Pt: 145…147 °C
Density: 1.3
Flash Pt: 46 °C
MDL Number: MFCD00042102
CAS Number: 350-57-2
UN: 1993
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca7b4a8c5103a98058318de8cc3178a31c7e3ebe&vid=ca7b4a8c5103a98058318de8cc3178a31c7e3ebe&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Boiling point 145-147°
Density 1.3
ND20 1.422

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty