Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methoxy-2,2,2-trifluoroethanol, Technical Grade

Supplier: MATRIX SCIENTIFIC MS

002901-25G 002901-100G
101776-416EA 280 USD InStock
101776-416 101776-418
1-Methoxy-2,2,2-trifluoroethanol, Technical Grade
1-Methoxy-2,2,2-trifluoroethanol
TCA listed: T
Hazard: Flammable


Formula: C₃H₅F₃O₂
Density: 1.36
MDL Number: MFCD00013572
CAS Number: 431-46-9
UN: 3271
ADR: 3,III
Order Now
Specification Test Results
ND20 1.3304

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty