Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2,2,3,3-tetrafluoropropionic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

003481-1G 003481-5G
101776-748EA 56.92 USD InStock
101776-748 101776-750
3-Chloro-2,2,3,3-tetrafluoropropionic acid ≥97%
3-Chloro-2,2,3,3-tetrafluoropropionic acid
Hazard: Corrosive


Formula: C₃HClF₄O₂
Boiling Pt: 130-131°C
Density: 1.63
MDL Number: MFCD00153099
CAS Number: 661-82-5
Order Now
Specification Test Results
Boiling point 130-131°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty