Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Diphenylpropiononitrile ≥97%

Supplier: MATRIX SCIENTIFIC MS

004720-25G 004720-100G
101777-704EA 76.92 USD InStock
101777-704 101777-706
2,2-Diphenylpropiononitrile ≥97%
2,2-Diphenylpropiononitrile
Hazard: Toxic


Formula: (C₆H₅)₂C(CN)CH₃
Boiling Pt: 145°C/2mm
Density: 1.109
MDL Number: MFCD00001846
CAS Number: 5558-67-8
UN: 3276
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e27ce7e18aa15b0826d9325a7b9518b82352b767&vid=e27ce7e18aa15b0826d9325a7b9518b82352b767&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Boiling point 145°/2mm
Density 1.109
ND20 1.572

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty