Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-4-(trifluoromethyl)benzonitrile ≥97%

Supplier: MATRIX SCIENTIFIC MS

005057-250MG 005057-1G
101777-924EA 118.46 USD InStock
101777-924 101777-926
2,6-Dichloro-4-(trifluoromethyl)benzonitrile ≥97%
2,6-Dichloro-4-(trifluoromethyl)benzonitrile
Hazard: Toxic


Formula: C₈H₂Cl₂F₃N
Boiling Pt: 66°C/1mm
MDL Number: MFCD00220879
CAS Number: 157021-61-9
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 66°/1mm

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty