Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Trifluoromethylbutyric acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

006029-1G 006029-5G
101778-834EA 58.46 USD InStock
101778-834 101778-836
3-Trifluoromethylbutyric acid ≥95%
3-Trifluoromethylbutyric acid
Hazard: Corrosive/Stench


Formula: C₅H₇F₃O₂
Boiling Pt: 90…91 °C (25 mmHg)
MDL Number: MFCD00039533
CAS Number: 348-75-4
Order Now
Specification Test Results
Boiling point 90-91°/25m
ND20 1.36

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty