Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-(trifluoromethyl)pyridine ≥98%

Supplier: MATRIX SCIENTIFIC MS

024615-25G 024615-5G
101780-386EA 67.69 USD InStock
101780-386 101780-384
2-Amino-3-(trifluoromethyl)pyridine ≥98%
2-Amino-3-(trifluoromethyl)pyridine
Hazard: Toxic/Irritant


Formula: C₆H₅F₃N₂
Melting Pt: 70…72 °C
MDL Number: MFCD04972538
CAS Number: 183610-70-0
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Melting point 70-72°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty