Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-FIH-1/HIF-1AN Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-428 NB100-428SS
102109-148EA 503.38 USD InStock
102109-148 102109-162
Anti-FIH-1/HIF-1AN Rabbit Polyclonal Antibody
Antibodies
The FIH-1 / HIF-1AN Antibody from Novus Biologicals is a rabbit polyclonal antibody to FIH-1 / HIF-1AN. This antibody reacts with human, mouse, rat. The FIH-1 / HIF-1AN Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy.

Type: Primary
Antigen: FIH-1/HIF-1AN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty