Electropolished Stainless Steel Cleanroom Tables, Solid Top, Eagle MHC™
Supplier: Eagle Group
SEP2436T SEP2448T SEP2460T SEP2472T SEP2484T SEP3036T SEP3048T SEP3060T SEP3072T SEP3084T SEP3636T SEP3648T SEP3660T SEP3672T SEP3684T
58202-756EA 4162.3 USD
58202-756 58202-758 58202-760 58202-762 58202-764 58202-768 58202-770 58202-772 58202-774 58202-776 58202-780 58202-782 58202-784 58202-786 58202-788
Electropolished Stainless Steel Cleanroom Tables, Solid Top, Eagle MHC™
Furniture Tables and Workbenches
14-gauge, Type 304 electropolished solid stainless top or brushed stainless steel and reinforced with heavy gauge hat channels.
  • Knocked down stainless steel C-frame
  • Tables supplied with 1" (25 mm) adjustable stainless steel bullet feet
  • Ends turned down 90° at 1¹⁄₂" (38 mm) providing for flush installations when required
Legs and crossbracing are 16 gauge 1⁵⁄₈" (41 mm) diameter with welded crossbracing front to back and choice of center or rear knocked-down crossbracing. Undershelves are heavy gauge construction with hat channel reinforcement on tables 84" (2134 mm) and longer.
Base assembly incorporates the 'strength by design' patented uni-lok® (patent #5,165,349) stainless steel gusset system which is welded into hat channel, reducing lateral movement.
Load capacity: 600 lbs.
Información para pedidos: Alcohol wiped and cleanroom bagged. Shipped knocked top reducing freight costs and potential for freight damage. Other base frame options available; contact your Avantor sales representative.
Order Now