Anti-IgG Goat Polyclonal Antibody (ATTO 488)

Supplier: Rockland Immunochemical
610-152-121 610-152-121S
RL610-152-121EA 436.72 USD
RL610-152-121 RL610-152-121S
Anti-IgG Goat Polyclonal Antibody (ATTO 488)
Antibodies
The emission spectra for this ATTO conjugate matches the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: ATTO 488
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR