Pentacopper silicide 99,5 Metals basis

Supplier: Origen Biomedical, Inc
Warning

012844-22 012844-14
AA12844-22EA 256 USD
AA12844-22 AA12844-14
Pentacopper silicide 99,5 Metals basis
Pentacopper silicide

Purity: 99.5% CAS No.: 12159-07-8 Molecular Formula: Cu5Si Formula Weight: 345.79 Physical Form: 6.35mm & Down Pieces Odor: Odorless MDL No.: MFCD00134719

Formula: Cu₅Si
MW: 345.79 g/mol
Melting Pt: 825 °C
Storage Temperature: Ambient
MDL Number: MFCD00134719
CAS Number: 12159-07-8
EINECS: 235-286-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR