Calcium molybdate ≥99%

Supplier: Origen Biomedical, Inc
Warning

043686-14 043686-22
AA43686-14EA 41.6 USD
AA43686-14 AA43686-22
Calcium molybdate ≥99%
Calcium molybdate

Purity: 99% CAS No.: 7789-82-4 Molecular Formula: CaMoO4 Formula Weight: 200.01 Physical Form: Powder Density: 4.45 Appearance: White Odor: Odorless Melting Point: 965° MDL No.: MFCD00015976

Formula: CaMoO₄
MW: 200.024 g/mol
Melting Pt: 965 °C
Density: 4.45
Storage Temperature: Ambient
MDL Number: MFCD00015976
CAS Number: 7789-82-4
EINECS: 232-192-9
Merck Index: 14,1681

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR