2-Bromoindene 98%
Supplier: Origen Biomedical, Inc
|
Warning
|
B25474-14
B25474-06
AAB25474-14EA
158
USD
AAB25474-14
AAB25474-06
AAB25474-03
2-Bromoindene 98%
2-Bromoindene
Purity: 98%
CAS No.: 10485-09-3
Molecular Formula: C9H7Br
Formula Weight: 195.06
Melting Point: 35-37°
MDL No.: MFCD06797863
Formula:
C₉H₇Br MW: 195.06 g/mol Melting Pt: 35…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797863 CAS Number: 10485-09-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...