(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) 97%

Supplier: Origen Biomedical, Inc
Warning

H26241-06 H26241-03
AAH26241-06EA 295 USD
AAH26241-06 AAH26241-03
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) 97%
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)

Purity: 97% CAS No.: 161265-03-8 Molecular Formula: C39H32OP2 Formula Weight: 578.62 Melting Point: 224-228° MDL No.: MFCD00233866

Formula: C₃₉H₃₂OP₂
MW: 578.62 g/mol
Melting Pt: 224…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00233866
CAS Number: 161265-03-8
EINECS: 605-249-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR