Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-methoxybenzylamine 97%

Supplier: Origen Biomedical, Inc
Danger

H26796-03 H26796-06
AAH26796-03EA 134 USD InStock
AAH26796-03 AAH26796-06
4-Fluoro-2-methoxybenzylamine 97%
4-Fluoro-2-methoxybenzylamine
Purity: 97% CAS No.: 870563-60-3 Molecular Formula: C8H10FNO Formula Weight: 155.17 Refractive Index: 1.5240


Formula: C₈H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD04116361
CAS Number: 870563-60-3
UN: 2735
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty