L05984-06
L05984-14
AAAL05984-06EA
16.6
USD
InStock
AAAL05984-06
AAAL05984-14
Phthalide-3-acetic acid ≥98%
Phthalide-3-acetic acid
Purity: 98+%
CAS No.: 4743-58-2
Molecular Formula: C10H8O4
Formula Weight: 192.17
Melting Point: 148-153°
Beilstein Reference: 156005
MDL No.: MFCD00051729