Calcium nitrate hydrate ≥99.9995% (metals basis), Puratronic®
Supplier: Origen Biomedical, Inc
|
Danger
|
044515-06
044515-14
044515-22
AA44515-06EA
55.37
USD
AA44515-06
AA44515-14
AA44515-22
Calcium nitrate hydrate ≥99.9995% (metals basis), Puratronic®
Calcium nitrate hydrate
Fieser: 18,84
Purity: 99.9995%
CAS No.: 35054-52-5
Molecular Formula: CaN2O6.xH2O
Formula Weight: 164.09(anhy)
Physical Form: Crystalline
Density: 1.82
Appearance: White
Melting Point: 42°
Boiling Point: 132° dec.
Merck Reference: 14,1682
UN No.: 1454
MDL No.: MFCD00149604
Formula:
Ca(NO₃)₂·H₂O MW: 182.1 g/mol Boiling Pt: 132 °C (dec.) Melting Pt: 42 °C Density: 1.82 Storage Temperature: Ambient |
MDL Number:
MFCD00149604 CAS Number: 35054-52-5 EINECS: 233-332-1 UN: 1454 ADR: 5.1 III Merck Index: 14,1682 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...