N-Ethylrhodanine 98%

Supplier: Origen Biomedical, Inc
Warning

B23047-14 B23047-06
AAB23047-14EA 261 USD
AAB23047-14 AAB23047-06
N-Ethylrhodanine 98%
N-Ethylrhodanine

Purity: 98% CAS No.: 7648-01-3 Molecular Formula: C5H7NOS2 Formula Weight: 161.25 Flash Point: 110°(230°F) Melting Point: 36-39° Boiling Point: 127-129°/4mm MDL No.: MFCD00051339

Formula: C₅H₇NOS₂
MW: 161.25 g/mol
Boiling Pt: 127…129 °C/4 mm
Melting Pt: 36…39 °C
Density: 1.303
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051339
CAS Number: 7648-01-3
EINECS: 231-603-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR