N-Acetylglycinamide(2-acetamidoacetamide) 97%

Supplier: Origen Biomedical, Inc

B24940-03 B24940-06 B24940-14
AAAB24940-03EA 50.2 USD
AAAB24940-03 AAAB24940-06 AAAB24940-14
N-Acetylglycinamide(2-acetamidoacetamide) 97%
N-Acetylglycinamide(2-acetamidoacetamide)

Purity: 97% CAS No.: 2620-63-5 Molecular Formula: CH3CONHCH2CONH2 Formula Weight: 116.12 Odor: Odorless Melting Point: 136-140° Beilstein Reference: 1753982 MDL No.: MFCD00008029

Formula: C₄H₈N₂O₂
MW: 116.12 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00008029
CAS Number: 2620-63-5
EINECS: 220-058-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR